DURAX™ Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78149-0 78149-00 78149-0C11 78149-1 78149-2 78149-2A 78149-3 78149-3C 78149-4 78149-5 78149-5A 78149-7 78149-7E 78149-F 78149-SS 78149-0C
102098-908EA 55.31 USD
102098-908 102098-910 102098-914 102098-916 102098-918 102098-920 102098-922 102098-924 102098-926 102098-928 102098-930 102098-932 102098-934 102098-936 102098-938 76376-126
DURAX™ Tweezers, Electron Microscopy Sciences
Forceps
With a 616HV Vickers-hardness, the tip of Rubis DURAX™ tweezers is six times harder than the tip of a standard steel tweezers.
  • Higher resistance against normal wear and tear
  • Longer service life
Even with intensive use, these tweezers retain their original precision. Their specially-hardened chrome-magnesium steel is also used in the manufacture of surgicals.