EMS Swiss Line Standard Tweezer Style 7, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
7801-7
76376-084EA 65.13 USD
76376-084
EMS Swiss Line Standard Tweezer Style 7, Electron Microscopy Sciences
Forceps
Swiss made standard tweezers have ultra fine pointed, curved, and angled precision tips with edges on the outside rounded off.