Phenol Red Broth with Galactose
Supplier: HARDY DIAGNOSTICS
Y306
76375-808PK 748.44 USD
76375-808
Phenol Red Broth with Galactose
Media for Microbiology Prepared Media for Microbiology
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.