VWR® PBS (Phosphate Buffered Saline) Buffer Powder Packs
Supplier: VWR International
EB1200
76371-734CS 147.99 USD
76371-734
VWR® PBS (Phosphate Buffered Saline) Buffer Powder Packs
Buffers Electrophoresis Buffers Washing Buffers
VWR® Buffer Powder Packs can save time and money, as there is no need to measure and weight many separate chemical components which is required when making buffers from scratch.
  • Buffer powder quickly dissolves to make 1L of 1X solution
  • No weighing or measuring of separate components
  • No pH adjustments required
  • Economical shipping, long shelf life
  • Quality and consistency guaranteed
VWR® Pre-measured buffer packs are extremely easy to use and are formulated so each pack makes 1 Liter of 1X buffer. Simply tear open a pack and mix completely with 1 liter of distilled water.
Shipping costs are also minimal, as each case of 50 or 100 packs is compact and weighs only 2.5 Lbs.
Certifications: COA avaialble for each lot number
Packaging: Each case of buffer packs is packaged in a convenient dispenser box, which takes up very little lab space. Included in each case is an instruction insert, and a roll of stickers to label each container of prepared buffer solution.