Zero Oxygen Solutions, Oakton®, Cole Parmer
Supplier: Antyila Scientific
00653-00
76371-706EA 29.7 USD
76371-706 76371-708
Zero Oxygen Solutions, Oakton®, Cole Parmer
Standards Environmental Analysis Standards
Solutions to maximize meter accuracy.
  • Ensure accuracy—tested using Winkler method for better results
  • Manufactured according to the APHA and ASTM formulation with high-purity, oxygen-free water