Overpack Poly Drums, Eagle Mfg
Supplier: Eagle Manufacturing
1661
76369-708EA 371.57 USD
76369-708
Overpack Poly Drums, Eagle Mfg
Barrels, Kegs, Drums
These overpack drums are constructed of blow-molded HDPE and are durable and lightweight.
  • Chemical and weather resistant
  • UV protection
  • Approved for use with packing groups i, ii and iii
Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. For use with larger non damaged containers, cans and drums of non leaking hazardous materials.
Certifications: UN certified. UL rating. Food grade quality - FDA 21 CFR 177.1520.