UV Lamps for Millipore Lab Water Systems, Avidity Science™
Supplier: AVIDITY SCIENCE
TR-QUVLQSL01 TR-ZMQUVLP01 TR-ZLXUVLP01 TR-ZFA10UVM1 TR-ZFA10UV01
76339-460EA 1083.87 USD
76339-460 76339-464 76361-244 76361-260 76361-262
UV Lamps for Millipore Lab Water Systems, Avidity Science™
Lights UV Lights, Lamps and Bulbs
These UV lamps are compatible with several lab water systems.
Available for following Millipore Lab Water Units: MilliQ Academic, MilliQ Advantage A10, MilliQ Biocel, MilliQ Element, MilliQ Gradient, MilliQ Integral, MilliQ Reference, MilliQ Synthesis, Elix, Super-Q Plus.