Polytron® Disposable 12 mm Generator, PT-DA 12/2SYN-E123, Kinematica
Supplier: Kinematica
11030035
76360-414PK 1307.1 USD
76360-414
Polytron® Disposable 12 mm Generator, PT-DA 12/2SYN-E123, Kinematica
Homogenizers Dispersing Tools
Disposable/reusable generators for when processing multiple samples per day, where the required sterilization of stainless steel generators represents a significant time limitation.
  • Prevention of cross-contamination
  • Standard dispersing applications