Digi-Sense™ Mini Particle Counter with Bluetooth® Connectivity, Cole-Parmer

Supplier: Argos Technologies
20250-65
76360-054EA 619.2 USD
76360-054
Digi-Sense™ Mini Particle Counter with Bluetooth® Connectivity, Cole-Parmer
Particle Counters
View, store, and share real-time data on your smartphone.

  • Get the most out of your data with the user-friendly mobile app
  • Unlimited data storage based on smart device capacity
  • Monitors particles, air temperature, and relative humidity
  • View measurements clearly on 2" color TFT LCD
  • Tilt stand offers easy hands-free viewing

Using the free Digi-Sense D/S Particle app, this mini particle counter with Bluetooth® technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your work—ideal for record keeping at large sites.

The handheld particle counter provides fast, easy, and accurate readings for particulate matter and particle mass concentration with air temperature and relative humidity measurements. It allows you to simultaneously measure PM2.5 (0 to 2000 ug/m³) and PM10 (0 to 2000 µg/m³). Other features include a selectable sample time, analog bar indicator, °F/°C selection, clock display, and auto power-off. Ideal for environmental protection and energy-saving applications.

Temperature resolution: 0.1 °C/°F

Humidity resolution: 0.1% RH

Memory: 5000 data sets and unlimited based on smart device capacity
Output: USB, Bluetooth®

Delivery information: Supplied with kick stand, protective cap, and one 9 V battery.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR