Digi-Sense™ Heavy-Duty Industrial Digital Multimeter with Bluetooth® Connectivity, Cole-Parmer
Supplier: Antyila Scientific
20250-64
76360-052EA 185.86 USD
76360-052
Digi-Sense™ Heavy-Duty Industrial Digital Multimeter with Bluetooth® Connectivity, Cole-Parmer
Electrical Meters Digital Multimeters
View, store, and share real-time data from a safe distance on your smartphone.
  • Get the most out of your data with our user-friendly mobile app
  • Unlimited data storage based on smart device capacity
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
  • Rugged double-molded housing provides increased durability
  • Bright, large backlit 4000-count LCD permits viewing in low-light conditions
  • Tilt stand offers easy hands-free viewing
  • IP67 waterproof rated
  • CAT IV-600 V, CAT III-1000 V safety rating
  • Continuity
  • Diode Tester
  • Duty Cycle
  • True Rms
Using the free Digi-Sense™ D/S Meter app, this heavy-duty industrial digital multimeter with Bluetooth® technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your work - ideal for record keeping at large sites.
The handheld digital multimeter measures AC/DC voltage, AC/DC current, resistance, frequency, capacitance, and continuity. Special functions include auto ranging, data Hold, Max/Min readings, diode test, audible continuity check, relative mode, duty cycle, low-battery indication, and auto power-off. The meter also offers True RMS for more accurate current readings, temperature measurement with a type K thermocouple, and analog bar graph for viewing trends.
Accessories information: Test leads, type K thermocouple probe with mini connector, carrying case, 9 V battery, and NIST-traceable calibration.