Digi-Sense™ AC/DC Clamp Meter with Bluetooth® Connectivity, Cole-Parmer
Supplier: Argos Technologies
20250-63
76360-050EA 247.8 USD
76360-050
Digi-Sense™ AC/DC Clamp Meter with Bluetooth® Connectivity, Cole-Parmer
Electrical Meters Digital Multimeters
View, store, and share real-time data on your smartphone.
  • Get the most out of your data with our user-friendly mobile app
  • Unlimited data storage based on smart device capacity
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
  • Rugged double-molded housing provides increased durability
  • Electronic overload protection prevents damage
  • Large backlit LCD permits viewing in low-light conditions
Using our free Digi-Sense D/S Meter app, this AC/DC clamp meter with Bluetooth® technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your work—ideal for record keeping at large sites.
The AC/DC clamp meter measures AC current to 1000 A AC, voltage (AC and DC), resistance, capacitance, frequency, temperature, duty cycle, and continuity. Special functions include auto ranging, data Hold, Max/Min readings, diode test, audible continuity check, over-range indication, low-battery indication, and auto power-off. The 1 3/16" (30 mm) jaw opening is ideal for use in small compartments or boxes with tightly arranged electrical wires. The meter also offers True RMS for more accurate current readings, and temperature measurement via a type K thermocouple.
The instrument is fully tested and calibrated to NIST-traceable standards for reliable measurements right out of the box, saving you time and money!
Certifications: NIST-traceable calibration report.
Delivery information: Supplied with test leads, type K thermocouple, carrying case, 9 V battery, and NIST-traceable calibration.