Oakton® Large-Dial Thermohygrometer, Cole-Parmer
Supplier: Antyila Scientific
35700-20
76360-046EA 107.22 USD
76360-046
Oakton® Large-Dial Thermohygrometer, Cole-Parmer
Hygrometers Analog Hygrometers
This large-dial thermohygrometer allows you to view temperature from a distance.
  • No batteries or power required
  • Big 10'' diameter
Ideal for the laboratory or greenroom, this indicator features a dual temperature display in °F and °C.
Membrane diameter: 10.75"
Depth: 1"