Cleaning Solution Pepsin-HCl, 250 ml
Supplier: Mettler Toledo
51350100
76359-838EA 39.98 USD
76359-838
Cleaning Solution Pepsin-HCl, 250 ml
Electrolytes Electrode Storage Solutions
Pepsin-HCl cleaning solution ideal for use with cleaning electrodes.
Keeps electrodes ready with the right cleaning, storage, and regeneration solutions.