Electrode Filling Solution, 3 M KCl Electrolyte
Supplier: Mettler Toledo
51350072
76359-830EA 28.34 USD
76359-830
Electrode Filling Solution, 3 M KCl Electrolyte
Electrolytes Electrode Filling Solutions
Mettler-Toledo offers electrolytes for every application. Fill-up your electrode with correct and fresh electrolyte for a precise measurement.
Sensor used: Reference cell, combined electrode
Parameter: pH and ISE