96-Square Well Magnetic Plates, Azer Scientific
Supplier: Azer Scientific Inc.
ES43001-0501 ES43001-0502 ES43001-0503
76359-802CS 520.79 USD
76359-802 76359-804 76359-806
96-Square Well Magnetic Plates, Azer Scientific
Microplates Deep Well Microplates
96 well magnetic plates and tip combs are designed for use in magnetic separation for DNA extraction and the filtration of proteins.
  • Slightly raised rims reduce the risk of contamination
  • Equipped with alphanumeric grid
The 96 square well magnetic plates and tip combs are made from high-purity polypropylene which has a high resistance to commonly used chemicals, solvents and alcohols. The plates and tip combs are designed for the filtration of proteins and DNA extractions through magnetic separation. The cylindrical wells enable a good bioturbation of samples and have a high stability during centrifugation.
Packaging: 60/case