Sapphire 48 Square Well Microplates, Azer Scientific
Supplier: Azer Scientific Inc.
ES43001-0062
76359-800CS 115.24 USD
76359-800
Sapphire 48 Square Well Microplates, Azer Scientific
Microplates Deep Well Microplates
Sapphire well plates meets SBS dimensional standards.
  • Square well, V-shaped bottom
  • Equipped with alphanumeric grid
  • Working volume up to 4.9 ml
The plates are manufactured out of pure polypropylene without lubricants for secure analysis. The thin-walled and parallel cavities ensure a maximum of contamination-free sample storage per well. Plates can be closed using cap mats, adhesive films and heat sealing.
Packaging: 20/case.