Glass Pipetting Syringe Assemblies, Cadence Science®
Supplier: Cadence Science
6700 6701 6703 6710 6711 6713 6720 6721 6723 6730 6731 6733
76359-600EA 794.7 USD
76359-600 76359-602 76359-604 76359-606 76359-608 76359-610 76359-612 76359-614 76359-616 76359-618 76359-620 76359-622
Glass Pipetting Syringe Assemblies, Cadence Science®
Syringes General-Purpose Syringes
Cadence Science® Glass pipetting syringes are equipped with contoured finger rings and thumb rest to allow for effortless, non-fatiguing one-handed operation.
  • Durable spring-loaded syringe assemblies
  • Automatic two-way valves
  • All units are autoclavable
Design features allow for self-refilling , which enables continuous pipetting from a single liquid reservoir. The pipettors are easily assembled. Ideal for use with sterile media during aseptic techniques.