M611, Labels for BMP®61 Labeling System BradyPrinter™, Brady
Supplier: Avantor
76359-224RL 152.56 USD
76359-224 76359-226 76359-228 76359-230 76359-232 76359-234 76359-236 76359-238 76359-240 76359-242 76359-244 76359-246 76359-248 76359-250 76359-252 76359-254 76359-256 76359-258 76359-260 76359-262 76359-264 76359-266 76359-268 76359-270 76359-272 76359-274 76359-276 76359-278 76359-280 76359-282 76359-284 76359-286 76359-288 76359-290 76359-294 76359-296 76359-298 76359-300 76359-302 76359-304 76359-306 76359-308 76359-310 76359-312 76359-314 76359-316 76359-318 76359-320 76359-322 76359-324 76359-326 76359-328 76359-330 76359-332 76359-334 76359-336 76359-338 76359-340 76359-342 76359-344 76359-346 76359-348 76359-350 76359-352 76359-354 76359-356 76359-358 76359-360 76359-362 76359-364 76359-366 76359-368 76359-370 76359-372 76359-374 76359-376 76359-378
M611, Labels for BMP®61 Labeling System BradyPrinter™, Brady
Labels
Keep data integrity and efficiency to complement current lab workflows with high performance identification labels.
  • An expanded laboratory material line for the BradyPrinter™ M611 to meet the most common lab applications
  • Long-lasting identification with materials designed to survive harsh lab conditions
Permanent Vinyl (B-439) is a thermal transfer printable material ideal for applications needing various colors like finished product ID, rating plates, and general-purpose IDs. Designed for use in ambient conditions with limited solvent exposure. Self-Laminating Polyester (B-461) is a clear film with a matte white printable zone ideal for cables, laboratory IDs, or general labeling where graphics need to be protected. Good smudge resistance, solvent resistance, and high/low-temperature performance.
Super chemical-resistant polyester (B-481) is designed for use in the slide staining process and other laboratory IDs which require superior chemical resistance. Performs well in the commonly used H&E (hematoxilin and eosin) slide staining process. Durable Polyester (B-490) markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws, and slides.
Durable Freezerbondz® Polyester (B-492) markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws, and slides. Permanent or repositionable (B-498) CleanLift® Series Vinyl Cloth, adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning.
Nylon cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as vials. Service temperature of –40 to 193 °F (–40 to 90 °C). Highly reflective B-584 material shines brightly when struck by light, which is ideal for applications where high visibility is critical.
Durable, low-shrink vinyl (B-595) with ultra-aggressive adhesive is the perfect solution for identification on rough surfaces and in harsh environment areas.