α-Hydroxy-o-tolyl-β-D-glucopyranoside

Supplier: RPI
Warning

S23200-100.0 S23200-500.0 S23200-1000.0
76348-204EA 431.22 USD
76348-204 76509-146 77510-436
α-Hydroxy-o-tolyl-β-D-glucopyranoside
α-Hydroxy-o-tolyl β-D-glucopyranoside

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₃H₁₈O₇
Melting Pt: 196...202 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00006590
CAS Number: 138-52-3
EINECS: 205-331-6

Order Now

Specification Test Results

Appearance White to off white powder
Identification Conforms to Structure
Purity (HPLC) ≥ 98.0 %
Loss on Drying (105°C, 4 hrs) ≤ 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR