Potassium chloride
Supplier: RPI
P41025-3000.0
P41025-500.0
P41025-1000.0
76347-760EA
166.41
USD
76347-760
76347-762
77510-398
Potassium chloride
Potassium chloride
Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
KCl Storage Temperature: Room Temperature |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 |
Specification Test Results
Appearance | Colorless crystals and/or white granular |
Assay | 99.0 - 100.5 % |
Barium | Pass Test |
Bromides | ≤ 0.010 % |
Calcium (Ca) | ≤ 0.002 % |
Chlorate and Nitrate (as NO3) | ≤ 0.003 % |
Heavy Metals (as Pb) | ≤ 5 ppm |
Insoluble Matter | ≤ 0.005 % |
Iodide (I) | ≤ 0.002 % |
Iron | ≤ 3 ppm |
Magnesium (Mg) | ≤ 0.001 % |
pH (5% Solution) | 5.4 - 8.6 |
Phosphate (PO4) | ≤ 5 ppm |
Sodium (Na) | ≤ 0.005 % |
Sulfate | ≤ 0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...