Potassium chloride

Supplier: RPI

P41025-3000.0 P41025-500.0 P41025-1000.0
76347-760EA 166.41 USD
76347-760 76347-762 77510-398
Potassium chloride
Potassium chloride

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: KCl
Storage Temperature: Room Temperature
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Appearance Colorless crystals and/or white granular
Assay 99.0 - 100.5 %
Barium Pass Test
Bromides ≤ 0.010 %
Calcium (Ca) ≤ 0.002 %
Chlorate and Nitrate (as NO3) ≤ 0.003 %
Heavy Metals (as Pb) ≤ 5 ppm
Insoluble Matter ≤ 0.005 %
Iodide (I) ≤ 0.002 %
Iron ≤ 3 ppm
Magnesium (Mg) ≤ 0.001 %
pH (5% Solution) 5.4 - 8.6
Phosphate (PO4) ≤ 5 ppm
Sodium (Na) ≤ 0.005 %
Sulfate ≤ 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR