Nitro blue tetrazolium chloride (NBT)

Supplier: RPI
Warning

N66000-0.1 N66000-1.0 N66000-5.0 N66000-0.5
76347-340EA 55.16 USD
76347-340 76347-342 76347-344 77510-364
Nitro blue tetrazolium chloride (NBT)
Nitro blue tetrazolium chloride (NBT)

Colorimetric substrate for dehydrogenases and other oxidases. NBT is used with X-Phos for colorimetric detection of alkaline phosphatase activity in blotting, immunohistochemical and cytochemistry techniques. Protect from light.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₄₀H₃₀Cl₂N₁₀O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00012159
CAS Number: 298-83-9
EINECS: 206-067-4

Order Now

Specification Test Results

Appearance Yellow Crystalline Powder
Identification Conforms
IR spectrum Concordant
Loss on Drying ≤ 9.5 %
Molar Absorptivity in Methyl Alcohol @257 nm ≥ 63000
Purity ≥ 98 %
Solubility Clear, Yellow Solution
Solubility (0.5% in H2O) Clear yellow solution
Solubility 1% in hot methanol Clear yellow solution
Sulfated Ash ≤ 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR