4-Methylumbelliferyl phosphate

Supplier: RPI

M63800-0.1 M63800-0.25 M63800-1.0 M63800-5.0
76346-926EA 94.9 USD
76346-926 76346-928 76346-930 77510-332
4-Methylumbelliferyl phosphate
4-Methylumbelliferyl phosphate

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₀H₉O₆P
Melting Pt: 215 °C
Storage Temperature: Freezer
MDL Number: MFCD00016969
CAS Number: 3368-04-5
EINECS: 222-137-7

Order Now

Specification Test Results

Appearance White to cream powder
Purity (TLC) ≥98%
Purity (Titration by NaOH) ≥98.0%
Solution Test Clear colorless to very pale yellow solution (1% H₂O)
Free 4-MU ≤100 ppm
Water Content (K.F) ≤0.5%
Identification FTIR Concordant
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR