Luria Broth Miller's Modified
Supplier: RPI
For propagation and maintenace of E.Coli.
- Low salt formula
- Molecular biology grade
Dissolve 15.5 grams in 1 liter distilled water and adjust the pH to 7.2. Sterilize by autoclaving for 15 minutes.
Caution: Research or further manufacturing use only, not for food or drug use.