Imidazole

Supplier: RPI
Danger

I52000-10.0 I52000-100.0 I52000-1000.0 I52000-500.0 I52000-5000.0 I52000-50.0
76346-306EA 34.46 USD
76346-306 76346-308 76346-310 76346-314 76346-316 77510-286
Imidazole
Imidazole

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₃H₄N₂
Storage Temperature: Room Temperature
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off white crystals
Assay ≥ 99.0 %
Identification Pass Test
Insolubles ≤ 0.1 %
Loss on Drying ≤ 0.5 %
Melting Point 87.0 to 91.0 oC
OPTICAL @ 260 NM ≤ 0.2
pH (5%, water) @25 oC 9.5 to 10.5
Solubility (20%, water) Pass
DNase Not Detected
Protease Not Detected
RNase Not Detected
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR