Guanidinium thiocyanate

Supplier: RPI
Danger

Synonyms: Guanidinium sulfocyanide, Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

G54000-100.0 G54000-1000.0 G54000-500.0 G54000-5000.0 G54000-250.0
76346-200EA 63.36 USD
76346-200 76346-202 76346-204 76346-206 77510-268
Guanidinium thiocyanate
Guanidinium thiocyanate

Protein denaturant used for purifying proteins and nucleic acids.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: CH₅N₃·CHNS
Storage Temperature: Room Temperature
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1

Order Now

Specification Test Results

Appearance White to slightly off white crystalline powder
Assay (Argentometric Titration) ≥ 99.0 %
Assay (Ion Chromatography) ≥ 99.5 %
Chloride Content ≤ 1.0 %
Cobalt (Co) ≤ 5 ppm
FTIR Conforms to reference
Iron ≤ 5 ppm
pH 5.0 - 7.0
DNase/ RNase None Detected
Melting Point 118 - 121 oC
Solubility Pass
Sulfate Ash ≤ 0.5 %
Moisture ≤ 0.5 %
BSE/TSE Free BSE/TSE Free
Grade Ultra Pure
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR