D-(+)-Glucose

Supplier: RPI

G32045-3000.0 G32045-500.0 G32045-10000.0
76346-100EA 141.04 USD
76346-100 76346-102 77510-266
D-(+)-Glucose
D(+)-Glucose

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₆H₁₂O₆
Storage Temperature: Room Temperature
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1

Order Now

Specification Test Results

Appearance White Powder
Chloride (Cl) ≤ 0.01 %
Heavy Metals ≤ 0.0005 %
Insoluble ≤ 0.005 %
Iron ≤ 0.0005 %
Loss on Drying (105 oC) ≤ 0.2 %
Residue after Ignition ≤ 0.02 %
Specific Rotation +52.5 - +53.0 °
Starch Pass Test
Sulfate and Sulfite (SO4) ≤ 0.005 %
Titrable Acid ≤ 0.002 meq/g
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR