Glycerine ACS

Supplier: RPI

G22025-0.5 G22025-20.0 G22025-4.0 G22025-10.0 G22025-1.0
76346-066EA 47.85 USD
76346-066 76346-068 76346-070 76509-166 77510-264
Glycerine ACS
Glycerine

Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Also used in casting gradient gels, as a protein stabilizer and storage buffer component.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: HOCH₂CH(OH)CH₂OH
Storage Temperature: Room Temperature
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5

Order Now

Specification Test Results

Appearance Clear liquid
Acrolein and Glucose Pass Test
Assay (GC) ≥ 99.5 %
Chlorinated compounds (as Cl) ≤ 0.003 %
Color, APHA ≤ 10
Fatty Acid Esters (as Butyric Acid) ≤ 0.05 %
Heavy Metals (as Pb) ≤ 2 ppm
Infrared Spectrum Conforms to standard
Neutrality Pass Test
Residue after Ignition ≤ 0.005 %
Substances Darkened By Sulfuric Acid Pass Test
Sulfate (SO4) ≤ 0.001 %
Water ≤ 0.5 %
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR