5-Fluoro-2'-deoxyuridine (floxuridine)

Supplier: RPI
Danger

Synonyms: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine

F10705-1.0 F10705-5.0
76345-984EA 305.18 USD
76345-984 77510-256
5-Fluoro-2'-deoxyuridine (floxuridine)
5-Fluoro-2'-deoxyuridine (Floxuridine)

Inhibits DNA synthesis by prior inhibition of thymidylate synthetase.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₉H₁₁FN₂O₅
Storage Temperature: Room Temperature
MDL Number: MFCD00006530
CAS Number: 50-91-9
EINECS: 200-072-5

Order Now

Specification Test Results

Appearance White or almost white crystalline powder
Assay (HPLC) ≥ 98.5 %
Fluoride ≤ 0.05 %
Heavy Metals (as Pb) ≤ 20 ppm
Identification Pass Test
Loss on Drying ≤ 0.5 %
Melting Range 145 - 153 °C
Residue on Ignition (Sulfated Ash) ≤ 0.1 %
Specific Rotation +36.0 - +39.0 °


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR