EDTA (ethylenediamine tetraacetic acid)

Supplier: RPI
Warning

E58100-500.0 E58100-1000.0 E58100-5000.0
76345-944EA 65.99 USD
76345-944 77510-252 77510-254
EDTA (ethylenediamine tetraacetic acid)
EDTA (Ethylenediamine tetraacetic acid)
Formula: C₁₀H₁₆N₂O₈
Storage Temperature: Room Temperature
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4

Order Now

Specification Test Results

Appearance Fine White Crystalline Powder
Assay (Titration) ≥ 99.0 %
Calcium (Ca) ≤ 0.001 %
Heavy Metals ≤ 0.001 %
Insoluble ≤ 1.0 %
Iron ≤ 0.005 %
Magnesium (Mg) ≤ 5 ppm
Grade ACS
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR