Cesium chloride, Optical Grade

Supplier: RPI
Warning

C68050-100.0 C68050-1000.0 C68050-250.0 C68050-500.0
76345-624EA 285.69 USD
76345-624 76345-626 77510-224 77510-226
Cesium chloride, Optical Grade
Cesium chloride

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: CsCl
Boiling Pt: 1,303 °C
Melting Pt: 645 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2

Order Now

Specification Test Results

Appearance Fine white powder
Aluminium (Al) ≤ 0.005 %
Barium (Ba) ≤ 0.005 %
Calcium (Ca) ≤ 0.005 %
Chromium (Cr) ≤ 1 ppm
Copper (Cu) ≤ 1 ppm
CsCl ≥ 99.9 %
Insoluble ≤ 100 ppm
Iron (Fe) ≤ 0.001 %
Lead (Pb) ≤ 0.001 %
Lithium (Li) ≤ 0.001 %
Magnesium (Mg) ≤ 0.001 %
Manganese (Mn) ≤ 1 ppm
Nickel (Ni) ≤ 1 ppm
Nuclease Activity None Detected
Optical Density @260 nm ≤ 0.02
Optical Density @405 nm ≤ 0.01
Particle Size 20 Mesh
Potassium (K) ≤ 0.005 %
Protease Activity None Detected
Rubidium (Rb) ≤ 0.02 %
Silicon (Si) ≤ 20 ppm
SO4 ≤ 0.01 %
Sodium (Na) ≤ 0.01 %
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR