Carbenicillin disodium salt ≥92.0% (by HPLC) USP

Supplier: RPI
Danger

C46000-1.0 C46000-100.0 C46000-25.0 C46000-5.0 C46000-500.0 C46000-50.0
76345-516EA 22.65 USD
76345-516 76345-518 76345-520 76345-522 76345-524 76345-526 77510-222
Carbenicillin disodium salt ≥92.0% (by HPLC) USP
Carbenicillin disodium salt

USP Grade. Prevents bacterial cell wall synthesis. Inhibits crosslinking of peptidoglycan.


  • Highly active against gram negative bacteria
  • Commonly used for the elimination of Agrobacterium species after inoculation
  • Sensitive to B-Lactamase
  • Non-toxic to plant cells
  • Plant cell culture tested
  • Hygroscopic, soluble in water

Formula: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
EINECS: 225-360-8

Order Now

Specification Test Results

Appearance White to Off White Crystalline Powder
Identification, Sodium Reaction Pass Test
Water ≤ 6.0 %
pH 6.50 - 8.0
Clarity of Solution Clear Solution
Benzyl penicillin sodium ≤ 5.0 %
Chromatographic Purity (HPLC) ≥ 92.0 %
Assay (Anhydrous) ≥ 88.0 %
Bacterial Endotoxins ≤ 0.05 EU/mg
Iodine Absorbing Substances ≤ 8.0 %
Potency (Dried basis) ≥ 770 µg/mg
Grade USP
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR