Calcium chloride dihydrate USP

Supplier: RPI
Warning

C36200-1000.0 C36200-500.0 C36200-5000.0
76345-432EA 96.64 USD
76345-432 76345-434 76345-436
Calcium chloride dihydrate USP
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Melting Pt: 176°C
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8

Order Now

Specification Test Results

Appearance White solid
Fluoride ≤ 0.004 %
Lead ≤ 5 mg/kg
Grade USP/FCC


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR