Citric acid monohydrate ACS

Supplier: RPI
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

C35025-1000.0 C35025-10000.0 C35025-500.0 C35025-5000.0 C35025-25000.0
76345-418EA 85.85 USD
76345-418 76345-420 76345-422 76345-424 77510-216
Citric acid monohydrate ACS
Citric acid monohydrate
Formula: C₆H₈O₇.H₂O
Boiling Pt: 175 °C
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1

Order Now

Specification Test Results

Appearance White crystalline powder
Phosphate ≤ 0.001 %
Purity 99.0 - 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR