Citric acid, anhydrous

Supplier: RPI
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

C34500-1000.0 C34500-500.0 C34500-5000.0 C34500-10000.0 C34500-25000.0
76345-410EA 65.99 USD
76345-410 76345-412 76345-414 77510-206 77510-208
Citric acid, anhydrous
Citric acid

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
Melting Pt: 153...159 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00011669
CAS Number: 77-92-9
Merck Index: 13,02350

Order Now

Specification Test Results

Appearance Fine white crystalline powder
Assay (Titration) ≥99.5 %
Carbonizable Substances (H2SO4) Pass Test
Clarity and Color Pass Test
Identification Pass Test
Lead ≤0.5 mg/kg
Limit of Oxalic Acid ≤0.036 %
Oxalate Test Pass Test
Residual Solvents Meet requirements
Residue on Ignition ≤0.05 %
Sulfate ≤0.015 %
Water ≤0.5 %
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR