D-(+)-Cellobiose

Supplier: RPI

Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

C32100-100.0 C32100-1000.0 C32100-25.0
76345-386EA 276.23 USD
76345-386 76345-388 76345-390
D-(+)-Cellobiose
D(+)-Cellobiose
Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00136034
CAS Number: 528-50-7
EINECS: 208-436-5

Order Now

Specification Test Results

Appearance White to slightly off white crystalline powder
Heavy Metals ≤ 20 ppm
Solubility Clear, colorless solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR