D-(+)-Cellobiose
Supplier: RPI
Synonyms:
β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
C32100-100.0
C32100-1000.0
C32100-25.0
76345-386EA
276.23
USD
76345-386
76345-388
76345-390
D-(+)-Cellobiose
D(+)-Cellobiose
Formula:
C₁₂H₂₂O₁₁ MW: 342.30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00136034 CAS Number: 528-50-7 EINECS: 208-436-5 |
Specification Test Results
Appearance | White to slightly off white crystalline powder |
Heavy Metals | ≤ 20 ppm |
Solubility | Clear, colorless solution |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...