Adonitol

Supplier: RPI

A32000-100.0 A32000-1000.0 A32000-2000.0 A32000-25.0 A32000-5.0 A32000-500.0
76344-886EA 412.86 USD
76344-886 76344-888 76344-890 76344-892 76344-894 76344-896
Adonitol
Adonitol

Adonitol is a crystalline pentose alcohol formed by the reduction of ribose.


  • It occurs naturally in the plant Adonis vernalis, as well as in the cell walls of Gram positive bacteria.

Formula: HOH₂C(CH(OH))₃CH₂OH
Storage Temperature: Ambient
MDL Number: MFCD00064291
CAS Number: 488-81-3
EINECS: 207-685-7

Order Now

Specification Test Results

Appearance White to off white powder
Identity (¹H NMR, D20) Conforms to structure
Solubility (100mg/2ml H2O) Colorless to faint yellow


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR