4-Aminobenzoic acid ≥98.5%

Supplier: RPI
Warning

A20050-100.0 A20050-25.0
76344-650EA 220.73 USD
76344-650 76344-652
4-Aminobenzoic acid ≥98.5%
4-Aminobenzoic acid

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₇H₇NO₂
Melting Pt: 180…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00007894
CAS Number: 150-13-0
EINECS: 205-753-0

Order Now

Specification Test Results

Appearance White crystalline powder
Assay ≥ 98.5 %
Loss on Drying ≤ 0.20 %
Melting Point 184 - 191 oC


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR