Agar 1 g, capsules, High Gel Strength

Supplier: RPI

A20021-1000.0 A20021-500.0
76344-626EA 533.66 USD
76344-626 76344-628
Agar 1 g, capsules, High Gel Strength
Agar

Each capsule contains 1g Agar.


  • Convenient and easy to use, simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring.

Melting Pt: 85…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Capsules (White to cream powder)
Ash ≤ 6.5 %
Gel Point 33 - 38 °C
Gel Strength (1.5% Solution @20 oC) 600 - 1100 g/cm²
Moisture ≤ 10 %
pH (1.5% solution) 6.0 - 8.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR