Adenosine 5'-monophosphate monohydrate

Supplier: RPI

A11240-1.0 A11240-10.0 A11240-5.0
76344-512EA 25.11 USD
76344-512 76344-514 76344-516
Adenosine 5'-monophosphate monohydrate
Adenosine-5'-monophosphoric acid (AMP) monohydrate

AMP [Adenosine-5-Monophosphate, Free Acid]


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₀H₁₄N₅O₇P·1H₂O
MDL Number: MFCD00149360
CAS Number: 18422-05-4
EINECS: 200-500-0

Order Now

Specification Test Results

Appearance Solid
Molar Absorptivity (@260nm - pH 7.5) 14.9 - 15.9 X103L/mole/cm
Ratio @ pH 7.5 A250/A260 0.75 - 0.81
Water Content ≤ 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR