N-Acetyl-L-cysteine ≥98.5%

Supplier: RPI

A10040-100.0 A10040-25.0 A10040-50.0 A10040-500.0 A10040-1000.0
76344-478EA 111.25 USD
76344-478 76344-480 76344-482 76344-484 77510-152
N-Acetyl-L-cysteine ≥98.5%
N-Acetyl-L-cysteine

Increases cellular pools of free radical scavenger


  • Prevents apoptotic death of neuronal cells.
  • Induces apoptosis in human and rat smooth muscle cells.
  • Pyrogen free.

Formula: C₅H₉NO₃S
Melting Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004880
CAS Number: 616-91-1
EINECS: 210-498-3

Order Now

Specification Test Results

Appearance White or almost crystalline powder
Loss on Drying ≤ 0.5 %
Heavy Metals (as Pb) ≤ 0.001 %
Assay 98.5 - 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR