VWR® Workstation Lab Mat

Supplier: VWR International
120785
76343-424EA 53.58 USD
76343-424
VWR® Workstation Lab Mat
Surface Protectors Bench Protectors
This durable FDA food grade silicone lab mat is ideal for every type of laboratory and more specialized areas such as cold rooms, cleanrooms, sterile suites and areas where high sterility is a requirement. They safely facilitate workflow processes with organization and easy clean up.

  • Reversible - Side one: Combination gridded and smooth surface; Side two: Smooth surface only
  • Keeps bench tops clean and safe from stains, spills and wear, protects against hot items up to 200 °C
  • Lip around the edge can hold approx. 250 ml of liquid
  • Anti-skid surface helps keep the mat and items in place
  • Noise dampening, reducing sound from the vibration of common bench top equipment
  • Chemically inert, does not react with most chemicals
  • Autoclavable at 121 °C for 15 minutes

The gridded surface is ideal for tube staging, preventing movement of small objects and securing items such as reservoirs, pipette tip boxes, assay plates, etc.

This mat is reusable and thus reduce waste making it a more environmentally friendly option over disposable mats. They can be easily cleaned using any standard disinfectant by simply wiping it and are dishwasher safe.

The mat can be rolled up for compact storage.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR