VWR® Parafilm® M Sealing Film, Purple
Supplier: VWR International
76343-422
76343-422CS 894.46 USD
76343-422
VWR® Parafilm® M Sealing Film, Purple
Seals General Purpose Sealing Films
Parafilm® M sealing film protects the integrity of your laboratory and research applications.
  • Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
  • Prevents moisture and volume loss due to material composition and tight seal
  • Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
  • Stretches more than 200% of original length
  • Disposable
Protect the integrity of research applications with a tight moisture-barrier that holds a strong seal and doesn't release from glassware, despite off-gassing.
Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions.