Memphis Welding Heavy-Duty Cowhide Leather Welding Blanket, MCR Safety
Supplier: MCR Safety
38063
76342-398EA 161.93 USD
76342-398
Memphis Welding Heavy-Duty Cowhide Leather Welding Blanket, MCR Safety
Welding Supplies
The memphis welding blanket is made from heavy side-split cow leather.
  • Natural pearl color
  • Heavy side-split leather construction
  • Stitched with DuPont™ Kevlar®
Featuring DuPont™ Kevlar® stitched seams that will resist tearing, sparks, and burn, the heavy-duty cowhide leather welding blanket provides outstanding protection from heat, spatter, sparks, slag, abrasion, and cuts.