Personna® Folding Utility Knife, AccuTec Blades, Inc.
Supplier: Accutec Blades
63-0221-0000
76339-884CS 338.49 USD
76339-884
Personna® Folding Utility Knife, AccuTec Blades, Inc.
Knives
Folding knife for convenient storage and safety.
  • Fixed blade
  • Quick blade change feature
  • Ergonomic, comfortable handle
Precision ground high carbon steel knife blades.