Hareta Wax Trimmer, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
HTR-PBT010
76339-792EA 814.86 USD
76339-792
Hareta Wax Trimmer, Springside Scientific
Paraffin Wax Trimmers and Dispensers
Used to remove excess wax from embedding tissue cassettes. Benchtop design fits easily in any lab. Melted paraffin easily drains into the removable tray with a convenient disposable tray liner.
  • 30% hotter and 30% faster than competitive products
  • Easy to use digital controls
  • Multiple blocks may be trimmed simultaneously
  • Grooved surface for easy trimming and wax drainage
  • Five disposable plastic inserts are provided
Temprature can be set between RT and 99.9° to meet specific application requirements. The device comes with a wax collection tray that adheres to the side of the unit with magnets.
Certifications: CE certified.