Monarch® gDNA Purification Columns, New England Biolabs
Supplier: New England Biolabs (NEB)
T3017L
76339-786EA 176 USD
76339-786
Monarch® gDNA Purification Columns, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Monarch® gDNA Purification Columns are a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010) and can be used to purify up to 30 µg of DNA from a wide variety of biological samples.
  • Designed without the use of a retaining ring, ensuring no buffer retention and no carryover of contaminants
DNA can be eluted in as little as 35 μl for and is ready for use in a variety of downstream applications including RT-PCR, RNA Library Prep for NGS, and transfection. Monarch® columns use less plastic than conventional columns, reducing their environmental footprint without compromising performance. They are supplied in quantities of 100, along with 200 Monarch® Collection Tubes II (NEB #T2018).