Monarch® gDNA Binding Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
T3014L
76339-780EA 144 USD
76339-780
Monarch® gDNA Binding Buffer, New England Biolabs
Buffers Extraction Buffers
The Monarch® gDNA Binding Buffer a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify up to 30 µg genomic DNA from a variety of sample type.
The Monarch® gDNA Binding Buffer is a chaotropic salt-based binding buffer with low alcohol content, designed to optimize binding of gDNA to the spin column. The binding of gDNA takes place at low speed, maximizing binding and efficiency.