Monarch® gDNA Blood Lysis Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
T3013L
76339-778EA 52 USD
76339-778
Monarch® gDNA Blood Lysis Buffer, New England Biolabs
Buffers Extraction Buffers
The Monarch® gDNA Blood Lysis Buffer a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify up to 30 µg genomic DNA from a variety of sample types.
The kit contains three unique lysis buffers, optimized to enable maximal yield and purity when preparing genomic DNA from a variety of sample types. The Blood Lysis Buffer contains a strong protective component against the high nuclease activity in blood samples and supports rapid degradation of hemoglobin and other protein components.
The Monarch® gDNA Blood Lysis Buffer is one of three lysis buffers available for purchase separately for additional convenience and to minimize waste.